Search
Search

faf| Gene Symbol: faf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260458.1 | NP_001247387.1 | 12 - 13 | 6843 | 73 | |
| NM_001260459.1 | NP_001247388.1 | 12 - 13 | 6891 | 73 | ||
| NM_001260460.2 | NP_001247389.2 | 12 - 13 | 6859 | 73 | ||
| NM_001260461.1 | NP_001247390.1 | 13 - 14 | 6957 | 73 | ||
| NM_001300698.1 | NP_001287627.1 | 12 - 13 | 6859 | 73 | ||
| NM_079873.5 | NP_524612.2 | 12 - 13 | 6859 | 73 | ||
| NM_170576.3 | NP_733455.1 | 12 - 13 | 6859 | 73 | ||
Set Membership: |
|