Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02152010_m1
          See other HEPH GE Assays ›
          Gene Symbol:
          heph
          Gene Name:
          hephaestus
          Gene Aliases:
          CG2094, CG2290, CG31000, DmPTB, Dmel\CG31000, Dmel_CG31000, HEPH, Heph, PTB, Ptb, dPTB, dmPTB, ema, hnRNP I, l(3)03429, l(3)j11B9, ptb
          Chromosome Location:
          Chr.3R: 31846987 - 32015520 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0011224
          Assay ID Dm02152010_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: heph
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104522.3 NP_001097992.1 5 - 6 611 65
          NM_001104523.3 NP_001097993.1 7 - 8 1431 65
          NM_001104524.3 NP_001097994.1 5 - 6 876 65
          NM_001170316.2 NP_001163787.1 7 - 8 1001 65
          NM_001170317.2 NP_001163788.1 5 - 6 599 65
          NM_001170319.1 NP_001163790.1 6 - 7 1039 65
          NM_001170320.2 NP_001163791.1 3 - 4 1258 65
          NM_001170321.2 NP_001163792.1 4 - 5 700 65
          NM_001170322.2 NP_001163793.1 4 - 5 1280 65
          NM_001170323.2 NP_001163794.1 6 - 7 1135 65
          NM_001260469.2 NP_001247398.1 7 - 8 1206 65
          NM_001260470.1 NP_001247399.1 8 - 9 1424 65
          NM_001300701.1 NP_001287630.1 5 - 6 1502 65
          NM_079964.3 NP_524703.1 3 - 4 522 65
          NM_170581.2 NP_733460.1 5 - 6 412 65
          NM_170582.4 NP_733461.2 5 - 6 876 65
          NM_176596.3 NP_788773.1 6 - 7 458 65
          NM_176597.3 NP_788774.1 5 - 6 1364 65
          NM_176598.2 NP_788775.1 5 - 6 970 65
          NM_176599.2 NP_788776.2 5 - 6 529 65
          NM_176600.3 NP_788777.1 4 - 5 569 65
          NM_176601.3 NP_788778.1 4 - 5 627 65
          NM_176602.3 NP_788779.1 4 - 5 389 65
          NM_176603.3 NP_788780.1 7 - 8 1001 65
          GenBank mRNA AF211191.2 - 3 - 4 544 65
          AF436844.1 - 7 - 8 1047 65
          AF455053.1 - 5 - 6 412 65
          AY052367.1 - 7 - 8 992 65
          BT030984.1 - 7 - 8 984 65
          BT032832.1 - 5 - 6 586 65

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Drosophila melanogaster

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          spermatid development
          sperm individualization
          negative regulation of oskar mRNA translation
          oocyte microtubule cytoskeleton polarization
          imaginal disc-derived wing vein morphogenesis
          imaginal disc-derived wing margin morphogenesis
          translation repressor activity, nucleic acid binding
          mRNA binding
          mRNA 3'-UTR binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline