Search
Search

Kat60| Gene Symbol: Kat60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275347.1 | NP_001262276.1 | 3 - 4 | 1073 | 64 | |
| NM_080258.4 | NP_524997.2 | 3 - 4 | 974 | 64 | ||
| GenBank mRNA | BT133173.1 | - | 3 - 4 | 983 | 64 | |
Set Membership: |
|