Search
Search

Atg16| Gene Symbol: Atg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144652.2 | NP_001138124.2 | 8 - 9 | 1853 | 88 | |
| NM_170432.3 | NP_733311.2 | 6 - 7 | 1138 | 88 | ||
| NM_170433.3 | NP_733312.2 | 5 - 6 | 1147 | 88 | ||
| NM_170434.4 | NP_733313.2 | 8 - 9 | 1829 | 88 | ||
| GenBank mRNA | AY058742.1 | - | 6 - 7 | 1138 | 88 | |
| BT024267.1 | - | 8 - 9 | 1779 | 88 | ||
Set Membership: |
|