Search
Search

Ppn| Gene Symbol: Ppn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170289.2 | NP_001163760.1 | 10 - 11 | 6625 | 85 | |
| NM_001170290.2 | NP_001163761.1 | 15 - 16 | 8626 | 85 | ||
| NM_176574.3 | NP_788751.2 | 14 - 15 | 8431 | 85 | ||
| NM_176575.3 | NP_788752.2 | 16 - 17 | 8797 | 85 | ||
Set Membership: |
|