Search
Search

AhcyL2| Gene Symbol: AhcyL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206499.2 | NP_996221.1 | 4 - 5 | 1133 | 101 | |
| NM_206500.2 | NP_996222.1 | 4 - 5 | 1010 | 101 | ||
| GenBank mRNA | AY113501.1 | - | 4 - 5 | 1007 | 101 | |
Set Membership: |
|