Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02154337_g1
          See other P53 GE Assays ›
          Gene Symbol:
          p53
          Gene Name:
          CG33336-PA
          Gene Aliases:
          CG10873, CG31325, CG33336, D-p53, DMP53, Dm-P53, DmP53, Dmel\CG33336, Dmel_CG33336, Dmp53, Dp53, P53, dmP53, dmp53, dp53, prac
          Chromosome Location:
          Chr.3R: 23049657 - 23054082 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0039044
          Assay ID Dm02154337_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: p53
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170223.1 NP_001163694.1 8 - 9 1768 74
          NM_001260323.2 NP_001247252.2 4 - 5 997 74
          NM_206544.2 NP_996267.1 8 - 9 1543 74
          NM_206545.2 NP_996268.1 7 - 8 1250 74
          GenBank mRNA AF192555.1 - 7 - 8 1234 74
          AF224713.1 - 7 - 8 1227 74
          AF224714.1 - 7 - 8 1234 74
          AF244918.1 - 7 - 8 1234 74
          AF250918.1 - 7 - 8 1234 74
          AF263722.1 - 7 - 8 1250 74
          AY069096.1 - 7 - 8 1234 74
          DQ191318.1 - 8 - 9 1483 74

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          P53-like transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of DNA repair
          regulation of transcription from RNA polymerase II promoter
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          reciprocal meiotic recombination
          neuroblast proliferation
          cell death
          determination of adult lifespan
          intrinsic apoptotic signaling pathway in response to DNA damage
          cellular response to starvation
          response to radiation
          response to UV
          response to ionizing radiation
          positive regulation of gene expression
          positive regulation of necrotic cell death
          neuroblast differentiation
          positive regulation of macroautophagy
          DNA damage response, signal transduction by p53 class mediator
          ectopic germ cell programmed cell death
          positive regulation of growth rate
          positive regulation of multicellular organism growth
          regulation of cell proliferation
          tissue regeneration
          positive regulation of protein import into nucleus
          response to starvation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of phosphatidylinositol 3-kinase activity
          positive regulation of insulin-like growth factor receptor signaling pathway
          positive regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of photoreceptor cell differentiation
          regulation of organ growth
          oogenesis
          regulation of apoptosis involved in tissue homeostasis
          cellular response to gamma radiation
          intrinsic apoptotic signaling pathway by p53 class mediator
          regulation of transcription from RNA polymerase II promoter in response to DNA damage
          positive regulation of cellular response to X-ray
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor binding
          TFIIH-class transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          promoter-specific chromatin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline