Search
Search

LpR1| Gene Symbol: LpR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104463.3 | NP_001097933.2 | 2 - 3 | 618 | 80 | |
| NM_001104464.2 | NP_001097934.2 | 2 - 3 | 618 | 80 | ||
| NM_001170263.2 | NP_001163734.1 | 2 - 3 | 618 | 80 | ||
| NM_001276043.1 | NP_001262972.1 | 2 - 3 | 618 | 80 | ||
| NM_001276044.1 | NP_001262973.1 | 3 - 4 | 2838 | 80 | ||
| NM_206573.2 | NP_996296.2 | 2 - 3 | 618 | 80 | ||
| GenBank mRNA | BT011057.1 | - | 2 - 3 | 618 | 80 | |
| BT011063.1 | - | 2 - 3 | 597 | 80 | ||
Set Membership: |
|