Search
Search

Inx3| Gene Symbol: Inx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276121.1 | NP_001263050.1 | 3 - 4 | 465 | 98 | |
| NM_001300646.1 | NP_001287575.1 | 3 - 4 | 512 | 98 | ||
| NM_079991.3 | NP_524730.1 | 3 - 4 | 465 | 98 | ||
| GenBank mRNA | AF172258.1 | - | 3 - 4 | 444 | 98 | |
| AY058643.1 | - | 3 - 4 | 438 | 98 | ||
Set Membership: |
|