Search
Search

vig2| Gene Symbol: vig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104443.2 | NP_001097913.1 | 2 - 3 | 523 | 145 | |
| NM_170204.2 | NP_733083.1 | 2 - 3 | 591 | 145 | ||
| NM_170205.3 | NP_733084.1 | 2 - 3 | 519 | 145 | ||
| NM_170206.3 | NP_733085.1 | 1 - 2 | 671 | 145 | ||
| GenBank mRNA | AY052117.1 | - | 1 - 2 | 638 | 145 | |
Set Membership: |
|