Search
Search

htl| Gene Symbol: htl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079670.3 | NP_524394.2 | 2 | 3758 | 129 | |
| NM_169784.2 | NP_732286.1 | 2 | 3531 | 129 | ||
| NM_169785.2 | NP_732287.1 | 2 | 3412 | 129 | ||
| GenBank mRNA | AY051812.1 | - | 2 | 3730 | 129 | |
Set Membership: |
|