Search
Search

Hsp60C| Gene Symbol: Hsp60C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135104.5 | NP_608948.2 | 3 | 1263 | 62 | |
| NM_164654.2 | NP_723104.2 | 3 | 1332 | 62 | ||
| NM_164655.3 | NP_723105.2 | 2 | 1199 | 62 | ||
| GenBank mRNA | BT001308.1 | - | 3 | 1209 | 62 | |
Set Membership: |
|