Search
Search

Mdh1| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298874.1 | NP_001285803.1 | 2 | 1091 | 84 | |
| NM_135550.2 | NP_609394.1 | 2 | 1034 | 84 | ||
| GenBank mRNA | BT128780.1 | - | 2 | 1001 | 84 | |
Set Membership: |
|