Search
Search

Chmp1| Gene Symbol: Chmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300179.1 | NP_001287108.1 | 2 | 665 | 85 | |
| NM_140794.3 | NP_649051.3 | 2 | 665 | 85 | ||
| GenBank mRNA | AY060798.1 | - | 2 | 620 | 85 | |
Set Membership: |
|