Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02371325_s1
          See other SKPA GE Assays ›
          Gene Symbol:
          SkpA
          Gene Name:
          SKP1-related A
          Gene Aliases:
          CG16983, Dmel\CG16983, Dmel_CG16983, EG:115C2.4, SKP1, SKPA, Skp, Skp1, anon-WO03040301.234, anon-WO03040301.236, anon-WO03040301.238, dSKP1, dSkip-1, dSkp-1, dSkpA, dskpA, l(1)G0037, l(1)G0058, l(1)G0109, l(1)G0389, skpA, spkA
          Chromosome Location:
          Chr.X: 656114 - 657899 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0025637
          Assay ID Dm02371325_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SkpA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038729.3 NP_001033818.1 1 1074 62
          NM_001297826.1 NP_001284755.1 1 356 62
          NM_058042.5 NP_477390.1 2 617 62
          NM_166856.3 NP_726690.1 2 465 62
          NM_166857.3 NP_726691.1 2 580 62
          NM_166858.3 NP_726692.1 2 1002 62
          NM_166859.3 NP_726693.1 2 399 62
          NM_166860.3 NP_726694.1 2 443 62
          NM_166861.3 NP_726695.1 2 375 62
          GenBank mRNA AY069297.1 - 1 1036 62
          BT099582.1 - 2 403 62
          BT099610.1 - 2 560 62
          KX532038.1 - 1 320 62

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          negative regulation of antimicrobial peptide production
          ubiquitin-dependent protein catabolic process
          neuron remodeling
          chromosome condensation
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          regulation of protein stability
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of hippo signaling
          DNA endoreduplication
          negative regulation of apoptotic process
          negative regulation of synaptic growth at neuromuscular junction
          positive regulation of mitotic cell cycle
          negative regulation of JNK cascade
          negative regulation of insulin receptor signaling pathway
          centrosome duplication
          negative regulation of peptidoglycan recognition protein signaling pathway
          negative regulation of canonical Wnt signaling pathway
          protein binding
          cullin family protein binding
          protein binding, bridging involved in substrate recognition for ubiquitination

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline