Search
Search

mRpL36| Gene Symbol: mRpL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144401.3 | NP_652658.1 | 2 | 166 | 70 | |
| GenBank mRNA | AY102667.1 | - | 2 | 147 | 70 | |
| KX531474.1 | - | 1 | 126 | 70 | ||
Set Membership: |
|