Search
Search

squ| Gene Symbol: squ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259115.2 | NP_001246044.1 | 2 | 769 | 71 | |
| NM_001273578.1 | NP_001260507.1 | 2 | 911 | 71 | ||
| NM_206000.4 | NP_995722.1 | 3 | 1112 | 71 | ||
| GenBank mRNA | BT089046.1 | - | 2 | 768 | 71 | |
Set Membership: |
|