Search
Search

RhoGAP1A| Gene Symbol: RhoGAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103385.2 | NP_001096855.1 | 9 - 10 | 3395 | 84 | |
| NM_001103386.2 | NP_001096856.1 | 9 - 10 | 3395 | 84 | ||
| NM_001169155.1 | NP_001162626.1 | 7 - 8 | 3597 | 84 | ||
| NM_001297795.1 | NP_001284724.1 | 9 - 10 | 3396 | 84 | ||
| NM_001297796.1 | NP_001284725.1 | 9 - 10 | 3402 | 84 | ||
| NM_001297797.1 | NP_001284726.1 | 7 - 8 | 3167 | 84 | ||
| GenBank mRNA | BT024449.1 | - | 9 - 10 | 3357 | 84 | |
| BT081989.1 | - | 7 - 8 | 3597 | 84 | ||
Set Membership: |
|