Search
Search

rl| Gene Symbol: rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015121.3 | NP_001015121.3 | 8 - 9 | 1201 | 113 | |
| NM_001015122.3 | NP_001015122.1 | 7 - 8 | 1132 | 113 | ||
| NM_001015123.3 | NP_001015123.1 | 6 - 7 | 1313 | 113 | ||
| NM_001110878.4 | NP_001104348.2 | 8 - 9 | 1170 | 113 | ||
| NM_001110879.2 | NP_001104349.1 | 7 - 8 | 1091 | 113 | ||
| NM_001300706.2 | NP_001287635.1 | 7 - 8 | 1108 | 113 | ||
| GenBank mRNA | AY070996.1 | - | 7 - 8 | 1132 | 113 | |
| M95124.1 | - | 6 - 7 | 1258 | 113 | ||
Set Membership: |
|