Search
Search

Gprk1| Gene Symbol: Gprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042973.3 | NP_001036438.1 | 12 - 13 | 1730 | 151 | |
| NM_001169580.2 | NP_001163051.1 | 13 - 14 | 1645 | 151 | ||
| NM_001299206.1 | NP_001286135.1 | 12 - 13 | 1730 | 151 | ||
| GenBank mRNA | BT022770.1 | - | 12 - 13 | 1663 | 151 | |
| BT022801.1 | - | 12 - 13 | 1705 | 151 | ||
Set Membership: |
|