Search
Search

nsfb| Gene Symbol: nsfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024454.2 | NP_001019625.2 | 5 - 6 | 452 | 135 | |
| NM_001354163.1 | NP_001341092.1 | 5 - 6 | 452 | 135 | ||
| XM_005156207.4 | XP_005156264.1 | 5 - 6 | 489 | 135 | ||
| XM_017358461.2 | XP_017213950.1 | 4 - 5 | 299 | 135 | ||
| GenBank mRNA | BC162793.1 | - | 5 - 6 | 430 | 135 | |
| BC162811.1 | - | 5 - 6 | 430 | 135 | ||
| DQ022212.1 | - | 5 - 6 | 452 | 135 | ||
Set Membership: |
|