Search
Search

g3bp1| Gene Symbol: g3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199956.1 | NP_956250.1 | 5 - 6 | 584 | 104 | |
| XM_005173102.4 | XP_005173159.1 | 5 - 6 | 624 | 104 | ||
| GenBank mRNA | BC045874.1 | - | 5 - 6 | 584 | 104 | |
| BC065323.1 | - | 5 - 6 | 535 | 104 | ||
Set Membership: |
|