Search
Search

hspa9| Gene Symbol: hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201326.2 | NP_958483.2 | 5 - 6 | 565 | 94 | |
| GenBank mRNA | BC044175.1 | - | 5 - 6 | 667 | 94 | |
| BC083504.1 | - | 5 - 6 | 565 | 94 | ||
| BC164566.1 | - | 5 - 6 | 575 | 94 | ||
Set Membership: |
|