Search
Search

prg4a| Gene Symbol: prg4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212753.2 | NP_997918.2 | 3 - 4 | 268 | 100 | |
| XM_005163364.4 | XP_005163421.1 | 3 - 4 | 291 | 100 | ||
| GenBank mRNA | BC050178.1 | - | 3 - 4 | 268 | 100 | |
Set Membership: |
|