Search
Search

gmppaa| Gene Symbol: gmppaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002196.1 | NP_001002196.1 | 9 - 10 | 1049 | 68 | |
| XM_005167476.4 | XP_005167533.1 | 10 - 11 | 1195 | 68 | ||
| GenBank mRNA | BC074036.1 | - | 9 - 10 | 1049 | 68 | |
Set Membership: |
|