Search
Search

acer1| Gene Symbol: acer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017603.1 | NP_001017603.1 | 6 - 7 | 800 | 88 | |
| XM_005166840.4 | XP_005166897.1 | 6 - 7 | 922 | 88 | ||
| GenBank mRNA | BC092849.1 | - | 6 - 7 | 800 | 88 | |
| BC164606.1 | - | 5 - 6 | 654 | 88 | ||
Set Membership: |
|