Search
Search

mmp23bb| Gene Symbol: mmp23bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017890.1 | NP_001017890.1 | 4 - 5 | 718 | 61 | |
| GenBank mRNA | BC092954.1 | - | 4 - 5 | 718 | 61 | |
| BC164476.1 | - | 4 - 5 | 569 | 61 | ||
Set Membership: |
|