Search
Search

mov10b.1| Gene Symbol: mov10b.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044342.2 | NP_001037807.2 | 10 - 11 | 1722 | 79 | |
| GenBank mRNA | BC171375.1 | - | 10 - 11 | 1722 | 79 | |
Set Membership: |
|