Search
Search

cgnb| Gene Symbol: cgnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044762.1 | NP_001038227.1 | 10 - 11 | 2257 | 121 | |
| XM_005159385.4 | XP_005159442.1 | 11 - 12 | 2455 | 121 | ||
| XM_005159386.4 | XP_005159443.1 | 10 - 11 | 2427 | 121 | ||
| GenBank mRNA | BC139880.1 | - | 10 - 11 | 2267 | 121 | |
| BC171708.1 | - | 12 - 13 | 2306 | 121 | ||
Set Membership: |
|