Search
Search

glsa| Gene Symbol: glsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045044.1 | NP_001038509.1 | 16 - 17 | 1641 | 64 | |
| XM_005161685.4 | XP_005161742.1 | 16 - 17 | 1703 | 64 | ||
| XM_021469341.1 | XP_021325016.1 | 10 - 11 | 4007 | 64 | ||
Set Membership: |
|