Search
Search

dclre1c| Gene Symbol: dclre1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045101.1 | NP_001038566.1 | 3 - 4 | 373 | 73 | |
| XM_017355061.2 | XP_017210550.1 | 7 - 8 | 888 | 73 | ||
| XM_021475059.1 | XP_021330734.1 | 6 - 7 | 1012 | 73 | ||
| XM_021475060.1 | XP_021330735.1 | 13 - 14 | 4246 | 73 | ||
| XM_021475061.1 | XP_021330736.1 | 12 - 13 | 4105 | 73 | ||
| XM_021475063.1 | XP_021330738.1 | 6 - 7 | 1435 | 73 | ||
| XM_021475064.1 | XP_021330739.1 | 5 - 6 | 852 | 73 | ||
| XM_021475065.1 | XP_021330740.1 | 4 - 5 | 887 | 73 | ||
| XM_021475066.1 | XP_021330741.1 | 7 - 8 | 887 | 73 | ||
| XM_021475067.1 | XP_021330742.1 | 7 - 8 | 899 | 73 | ||
Set Membership: |
|