Search
Search

plcd3a| Gene Symbol: plcd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099423.1 | NP_001092893.1 | 9 - 10 | 1868 | 72 | |
| GenBank mRNA | BC139849.1 | - | 9 - 10 | 1868 | 72 | |
Set Membership: |
|