Search
Search

tbx16l| Gene Symbol: tbx16l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131052.2 | NP_571127.2 | 7 - 8 | 789 | 78 | |
| GenBank mRNA | BC099989.1 | - | 7 - 8 | 762 | 78 | |
| U80951.1 | - | 7 - 8 | 798 | 78 | ||
Set Membership: |
|