Search
Search

cyp19a1b| Gene Symbol: cyp19a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131642.2 | NP_571717.2 | 5 - 6 | 721 | 111 | |
| XM_021470523.1 | XP_021326198.1 | 5 - 6 | 1081 | 111 | ||
| GenBank mRNA | AF183908.1 | - | 5 - 6 | 700 | 111 | |
| AF226619.1 | - | 5 - 6 | 704 | 111 | ||
Set Membership: |
|