Search
Search

atp1a2a| Gene Symbol: atp1a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131683.1 | NP_571758.1 | 9 - 10 | 1306 | 91 | |
| GenBank mRNA | AF286373.1 | - | 9 - 10 | 1306 | 91 | |
| AY028628.1 | - | 9 - 10 | 1240 | 91 | ||
| BC163629.1 | - | 9 - 10 | 1228 | 91 | ||
Set Membership: |
|