Search
Search

mkrn1| Gene Symbol: mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152978.1 | NP_694510.1 | 1 - 2 | 93 | 66 | |
| GenBank mRNA | AF277173.2 | - | 1 - 2 | 64 | 66 | |
| BC095243.1 | - | 1 - 2 | 93 | 66 | ||
| BC164736.1 | - | 1 - 2 | 114 | 66 | ||
Set Membership: |
|