Search
Search

cyp2k22| Gene Symbol: cyp2k22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200235.1 | NP_956529.1 | 4 - 5 | 723 | 74 | |
| GenBank mRNA | AF487990.1 | - | 4 - 5 | 718 | 74 | |
| BC046898.1 | - | 4 - 5 | 723 | 74 | ||
| BC065455.1 | - | 4 - 5 | 707 | 74 | ||
| BC154313.1 | - | 4 - 5 | 694 | 74 | ||
| BC164816.1 | - | 4 - 5 | 707 | 74 | ||
Set Membership: |
|