Search
Search

ercc2| Gene Symbol: ercc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200926.2 | NP_957220.2 | 17 - 18 | 1849 | 119 | |
| GenBank mRNA | BC049410.1 | - | 17 - 18 | 1759 | 119 | |
| JN172105.1 | - | 16 - 17 | 1766 | 119 | ||
Set Membership: |
|