Search
Search

cry-dash| Gene Symbol: cry-dash | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205686.1 | NP_991249.1 | 8 - 9 | 866 | 87 | |
| GenBank mRNA | AB120759.1 | - | 8 - 9 | 866 | 87 | |
| BC098514.1 | - | 8 - 9 | 936 | 87 | ||
Set Membership: |
|