Search
Search

tdh| Gene Symbol: tdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213245.1 | NP_998410.1 | 7 - 8 | 769 | 75 | |
| XM_005160456.2 | XP_005160513.1 | 7 - 8 | 883 | 75 | ||
| GenBank mRNA | BC063962.1 | - | 7 - 8 | 769 | 75 | |
Set Membership: |
|