Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: sulf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003833.2 | NP_001003833.2 | 7 - 8 | 1288 | 96 | |
XM_017353509.1 | XP_017208998.1 | 7 - 8 | 1298 | 96 | ||
XM_017353510.2 | XP_017208999.1 | 9 - 10 | 1601 | 96 | ||
XM_017353511.2 | XP_017209000.1 | 7 - 8 | 1202 | 96 | ||
XM_021469735.1 | XP_021325410.1 | 11 - 12 | 2020 | 96 | ||
XM_021469736.1 | XP_021325411.1 | 6 - 7 | 1068 | 96 | ||
XM_021469737.1 | XP_021325412.1 | 6 - 7 | 1068 | 96 | ||
XM_021469738.1 | XP_021325413.1 | 6 - 7 | 1068 | 96 | ||
XM_021469739.1 | XP_021325414.1 | 6 - 7 | 1068 | 96 | ||
GenBank mRNA | AY332607.1 | - | 7 - 8 | 1288 | 96 | |
AY332608.1 | - | 6 - 7 | 1068 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |