Search
Search

sf3a3| Gene Symbol: sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004289.1 | NP_001004289.1 | 6 - 7 | 593 | 91 | |
| GenBank mRNA | AY648757.1 | - | 6 - 7 | 593 | 91 | |
| BC092810.1 | - | 6 - 7 | 534 | 91 | ||
| BC096781.1 | - | 6 - 7 | 594 | 91 | ||
| BC165622.1 | - | 6 - 7 | 499 | 91 | ||
Set Membership: |
|