Search
Search

snx9a| Gene Symbol: snx9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012313.2 | NP_001012313.2 | 10 - 11 | 1215 | 93 | |
| XM_005158722.4 | XP_005158779.1 | 10 - 11 | 1215 | 93 | ||
| XM_005158723.4 | XP_005158780.1 | 10 - 11 | 1206 | 93 | ||
| GenBank mRNA | BC091825.1 | - | 10 - 11 | 1219 | 93 | |
| BC164273.1 | - | 9 - 10 | 1001 | 93 | ||
Set Membership: |
|