Search
Search

smyd2a| Gene Symbol: smyd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013550.1 | NP_001013568.1 | 1 - 2 | 315 | 65 | |
| GenBank mRNA | BC091465.1 | - | 1 - 2 | 315 | 65 | |
| DQ851823.1 | - | 1 - 2 | 198 | 65 | ||
Set Membership: |
|