Search
Search

pacsin1a| Gene Symbol: pacsin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328357.1 | NP_001315286.1 | 5 - 6 | 816 | 114 | |
| XM_005161905.4 | XP_005161962.1 | 5 - 6 | 796 | 114 | ||
| XM_021469807.1 | XP_021325482.1 | 5 - 6 | 797 | 114 | ||
| GenBank mRNA | BC095091.1 | - | 5 - 6 | 784 | 114 | |
| BC164592.1 | - | 4 - 5 | 631 | 114 | ||
Set Membership: |
|