Search
Search

slit1a| Gene Symbol: slit1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034968.1 | NP_001030140.1 | 31 - 32 | 3459 | 73 | |
| XM_009307022.3 | XP_009305297.1 | 28 - 29 | 3178 | 73 | ||
| GenBank mRNA | BC163538.1 | - | 31 - 32 | 3351 | 73 | |
| DQ177286.1 | - | 31 - 32 | 3459 | 73 | ||
Set Membership: |
|