Search
Search

cdk4| Gene Symbol: cdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077777.1 | NP_001071245.1 | 2 - 3 | 306 | 88 | |
| GenBank mRNA | BC124438.1 | - | 2 - 3 | 306 | 88 | |
| BC164244.1 | - | 1 - 2 | 249 | 88 | ||
Set Membership: |
|