Search
Search

adpgk| Gene Symbol: adpgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079965.2 | NP_001073434.1 | 3 - 4 | 617 | 60 | |
| XM_005162956.4 | XP_005163013.1 | 4 - 5 | 756 | 60 | ||
| XM_005162957.4 | XP_005163014.1 | 3 - 4 | 833 | 60 | ||
| GenBank mRNA | BC125924.1 | - | 3 - 4 | 551 | 60 | |
Set Membership: |
|