Search
Search

kiz| Gene Symbol: kiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098390.1 | NP_001091860.1 | 6 - 7 | 1228 | 86 | |
| XM_005158902.4 | XP_005158959.1 | 6 - 7 | 1302 | 86 | ||
| GenBank mRNA | BC141787.1 | - | 6 - 7 | 1228 | 86 | |
Set Membership: |
|