Search
Search

selenot2| Gene Symbol: selenot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098487.2 | NP_001091957.2 | 4 - 5 | 634 | 96 | |
| GenBank mRNA | BC065465.1 | - | 4 - 5 | 606 | 96 | |
| BC095658.1 | - | 4 - 5 | 599 | 96 | ||
Set Membership: |
|